EMAIL THIS PAGE TO A FRIEND
The Science of the Total Environment
The effects of different coatings on zinc oxide nanoparticles and their influence on dissolution and bioaccumulation by the green alga, C. reinhardtii.
PMID 24836387