EMAIL THIS PAGE TO A FRIEND
Leukemia research
Circulating YKL-40 in patients with essential thrombocythemia and polycythemia vera treated with the novel histone deacetylase inhibitor vorinostat.
PMID 24836761
Abstract
YKL-40 regulates vascular endothelial growth factors and induces tumor proliferation. We investigated YKL-40 before and after treatment with vorinostat in 31 polycythemia vera (PV) and 16 essential thrombocythemia (ET) patients. Baseline PV patient levels were 2 times higher than in healthy controls (P<0.0001) and 1.7 times higher than in ET (P=0.02). A significant correlation between YKL-40 at baseline and neutrophils, CRP, LDH, JAK2V617F and platelets in PV patients was observed, as well as a significantly greater reduction of YKL-40 levels in PV patients responding to therapy. YKL-40 might be a novel marker of disease burden and progression in myeloproliferative neoplasms.