EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Polyethylene glycol modified magnetic carbon nanotubes as nanosorbents for the determination of methylprednisolone in rat plasma by high performance liquid chromatography.
PMID 24837418