EMAIL THIS PAGE TO A FRIEND
Food chemistry
Investigating the potential of under-utilised plants from the Asteraceae family as a source of natural antimicrobial and antioxidant extracts.
PMID 24837924
Abstract
Antimicrobial properties of ethanol and water extracts from eight Asteraceae species were investigated against three Gram positive (Staphylococcus aureus, MRSA and Bacillus cereus) and two Gram negative (Escherichia coli and Salmonella typhimurium) bacterial strains. Ethanol extracts from Centaurea scabiosa, Arctium minus, Taraxacum officinale, Centaurea nigra and Cirsium palustre demonstrated antimicrobial activity against strains of S. aureus, MRSA and B. cereus (MIC=187.5-365μg/ml). Ethanol extracts also had higher antioxidant activities and phenolic content demonstrating a link between these compounds and the bioactivity of these extracts. Further investigation into the phenolic content of ethanol extracts using UPLC-MS/MS lead to the identification and quantification of numerous phenolic compounds in all species including; 18 from Cirsium arvense, 16 from Cirsium vulgare, 19 from C. palustre, 15 from C. nigra, 17 from C. scabiosa, 14 from Sonchus asper, 17 from A. minus and 11 from T. officinale.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
03910590
(±)-Catechin hydrate, primary reference standard
C15H14O6 · xH2O
43412
(+)-Catechin, analytical standard
C15H14O6
E1753
(−)-Epicatechin, ≥90% (HPLC)
C15H14O6
68097
(−)-Epicatechin, analytical standard
C15H14O6
E4018
(−)-Epicatechin, ≥98% (HPLC), from green tea
C15H14O6
E3893
(−)-Epicatechin gallate, ≥98% (HPLC), from green tea
C22H18O10
50299
(−)-Epigallocatechin gallate, ≥97.0% (HPLC)
C22H18O11
93894
(−)-Epigallocatechin gallate, analytical standard
C22H18O11
17779
2,3,5-Triphenyl-tetrazolium chloride solution, for microbiology
C19H15ClN4
93285
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0% (HPLC)
C18H12N6
T1253
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. (of Fe), ≥98%
C18H12N6
37580
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
C7H6O4
08992
3,4-Dihydroxybenzoic acid, analytical standard
C7H6O4
42251
Apigenin, analytical standard
C15H10O5
01760595
Apigenin, primary reference standard
C15H10O5
10798
Apigenin, ≥95.0% (HPLC)
C15H10O5
60018
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
C9H8O4
00500590
Chlorogenic acid, primary reference standard
C16H18O9
Y0000569
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
C16H18O9
138622
D-(−)-Quinic acid, 98%
C7H12O6
03940590
Epicatechin, primary reference standard
C15H14O6
03950590
Epicatechin gallate, primary reference standard
C22H18O10
03970590
Epigallocatechin gallate, primary reference standard
C22H18O11
PHR1333
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
C22H18O11
48640
Ethyl gallate, antioxidant, ≥96.0% (HPLC)
C9H10O5
Y0001013
Ferulic acid, European Pharmacopoeia (EP) Reference Standard
C10H10O4
I8377
Iodonitrotetrazolium chloride, Used in colorimetric assays.
C19H13ClIN5O2
58030
Iodonitrotetrazolium chloride, BioReagent, ≥97.0% (calc. on dry substance, NT)
C19H13ClIN5O2
I10406
Iodonitrotetrazolium chloride, 95%
C19H13ClIN5O2
96353
Kaempferol, analytical standard
C15H10O6