EMAIL THIS PAGE TO A FRIEND
Journal of Agricultural and Food Chemistry
Synthesis of caffeic acid phenethyl ester derivatives, and their cytoprotective and neuritogenic activities in PC12 cells.
PMID 24840770