EMAIL THIS PAGE TO A FRIEND
Experimental and Molecular Pathology
Detection of MYD88 L265P mutations in formalin-fixed and decalcified BM biopsies from patients with lymphoplasmacytic lymphoma.
PMID 24842316