EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Qualitative and quantitative determination of peptides related to celiac disease in mixtures derived from different methods of simulated gastrointestinal digestion of wheat products.
PMID 24842402
Abstract
During wheat digestion, gluten-derived proteolytic resistant peptides are generated, some of them involved in celiac disease. In vitro digestion models able to mimic the peptides generated in the human gastrointestinal tract are extremely useful to assess the pathogenicity of wheat-derived products. In this paper, samples belonging to three different durum wheat varieties were taken at six different steps of the pasta production chain and two different digestion models present in the literature were assessed on the different samples: a more complex one using artificial fluids simulating the exact composition of digestive juices, and a simplified method based on a peptic-tryptic/chymotryptic treatment of wheat ethanolic extract. An extensive characterization of the peptides generated using two in vitro digestion models was performed through LC-MS/MS techniques and the two methods were compared in order to evaluate qualitative and quantitative differences and their possible implications for varietal screening. Strong differences in the type of peptides produced with the two methods were detected, indicating that the simplified method can still be used for a varietal screening but is not representative of the peptides really generated after physiological human digestion. Results indicate a clear necessity of physiologically accurate models for simulating human gastrointestinal digestion of wheat products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45983
Acetonitrile, analytical standard
C2H3N
PHR1551
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
A9434
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
H4ClN
09718
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
H4ClN
254134
Ammonium chloride, 99.998% trace metals basis
H4ClN
326372
Ammonium chloride, 99.99% trace metals basis
H4ClN
PHR1327
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
H4ClN
09724
Ammonium chloride, tested according to Ph Eur
H4ClN
09683
Ammonium ion solution for ISE, 0.1 M NH4+, analytical standard (for ion-selective electrodes)
H4ClN
09685
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
H4ClN
21059
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
CaCl2
G5269
D-Glucuronic acid, ≥98% (GC)
C6H10O7
G4875
D-(+)-Glucosamine hydrochloride, ≥99%, crystalline
C6H13NO5 · HCl
G1514
D-(+)-Glucosamine hydrochloride, ≥99%, BioReagent, suitable for cell culture
C6H13NO5 · HCl
1012772
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
C2H6O
PHR1070
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
C2H6O
296082
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
C4H10O
91238
Diethyl ether, analytical standard
C4H10O
PHR1553
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO
78040
DL-Phenylalanine, puriss., ≥99.0% (NT)
C9H11NO2
W372609
DL-Phenylalanine, 99%, FCC
C9H11NO2
147966
DL-Phenylalanine, ReagentPlus®, 99%
C9H11NO2
145726
DL-Tyrosine, 99%
C9H11NO3
06473
Formic acid solution, BioUltra, 1.0 M in H2O
CH2O2
Y0001381
Glucosamine for system suitability, European Pharmacopoeia (EP) Reference Standard
C6H13NO5 · HCl
1294207
Glucosamine hydrochloride, United States Pharmacopeia (USP) Reference Standard
C6H13NO5 · HCl
PHR1199
Glucosamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C6H13NO5 · HCl
Y0001406
Glucosamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C6H13NO5 · HCl
12804
HBTU, ≥98.0% (T)
C11H16F6N5OP
387649
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
C8H19N