EMAIL THIS PAGE TO A FRIEND
Antioxidants & Redox Signaling
Hypoxia-responsive microRNA-101 promotes angiogenesis via heme oxygenase-1/vascular endothelial growth factor axis by targeting cullin 3.
PMID 24844779