EMAIL THIS PAGE TO A FRIEND
Annals of Hematology
High prognostic value of minimal residual disease detected by flow-cytometry-enhanced fluorescence in situ hybridization in core-binding factor acute myeloid leukemia (CBF-AML).
PMID 24844781