EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical Sciences
Intramuscular administration of paliperidone palmitate extended-release injectable microsuspension induces a subclinical inflammatory reaction modulating the pharmacokinetics in rats.
PMID 24845884