EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International Edition in English)
A graphene oxide membrane with highly selective molecular separation of aqueous organic solution.
PMID 24846755
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
642991
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Al2O3
702129
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Al2O3
05184
Aluminum oxide, for the determination of hydrocarbons
Al2O3
06300
Aluminum oxide, activated, neutral, Brockmann Activity I
Al2O3
67993
Aluminum oxide, for chromatography
Al2O3
544833
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Al2O3
551643
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Al2O3
202606
Aluminum oxide, 99.997% trace metals basis
Al2O3
634875
Aluminum oxide, single crystal substrate, <0001>
Al2O3
718475
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Al2O3
517747
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Al2O3
P2097
Potassium permanganate, meets USP testing specifications
KMnO4
238511
Potassium permanganate, ≤150 μm particle size, 97%
KMnO4
207985
Potassium permanganate, 97%
KMnO4
60459
Potassium permanganate, BioUltra, ≥99.0% (RT)
KMnO4
60458
Potassium permanganate, puriss. p.a., ACS reagent, Hg ≤0.000005%, ≥99.0% (RT)
KMnO4
31404
Potassium permanganate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
KMnO4
13206
Potassium permanganate, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
KMnO4
60467
Potassium permanganate, tested according to Ph.Eur.
KMnO4
HT1022
Potassium permanganate solution
KMnO4