PloS one
Variations of CITED2 are associated with congenital heart disease (CHD) in Chinese population.
PMID 24848765
Abstract
CITED2 was identified as a cardiac transcription factor which is essential to the heart development. Cited2-deficient mice showed cardiac malformations, adrenal agenesis and neural crest defects. To explore the potential impact of mutations in CITED2 on congenital heart disease (CHD) in humans, we screened the coding region of CITED2 in a total of 700 Chinese people with congenital heart disease and 250 healthy individuals as controls. We found five potential disease-causing mutations, p.P140S, p.S183L, p.S196G, p.Ser161delAGC and p. Ser192_Gly193delAGCGGC. Two mammalian two-hybrid assays showed that the last four mutations significantly affected the interaction between p300CH1 and CITED2 or HIF1A. Further studies showed that four CITED2 mutations recovered the promoter activity of VEGF by decreasing its competitiveness with HIF1A for binding to p300CH1 and three mutations decreased the consociation of TFAP2C and CITED2 in the transactivation of PITX2C. Both VEGF and PITX2C play very important roles in cardiac development. In conclusion, we demonstrated that CITED2 has a potential causative impact on congenital heart disease.