EMAIL THIS PAGE TO A FRIEND
Organic letters
Visible-light-mediated fluoroalkylation of isocyanides with ethyl bromofluoroacetates: unified synthesis of mono- and difluoromethylated phenanthridine derivatives.
PMID 24850129
Abstract
A practical and unified strategy has been described for the preparation of mono- and difluoromethylated phenanthridine derivatives using a visible-light-promoted alkylation and decarboxylation sequence from biphenyl isocyanides with ethyl bromofluoroacetate (EBFA) or ethyl bromodifluoroacetate (EBDFA). These reactions could be carried out at room temperature in good to excellent chemical yields. Both stepwise and one-pot procedures have been developed, which makes this strategy more attractive.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295795
Ethyl bromodifluoroacetate, 98%
C4H5BrF2O2
452114
Ethyl bromofluoroacetate, 97%
C4H6BrFO2