EMAIL THIS PAGE TO A FRIEND
Clinical cancer research : an official journal of the American Association for Cancer Research
Real-time immune monitoring to guide plasmid DNA vaccination schedule targeting prostatic acid phosphatase in patients with castration-resistant prostate cancer.
PMID 24850844
Abstract
We have previously reported that a DNA vaccine encoding prostatic acid phosphatase (PAP) could elicit PAP-specific T cells in patients with early recurrent prostate cancer. In the current pilot trial, we sought to evaluate whether prolonged immunization with regular booster immunizations, or "personalized" schedules of immunization determined using real-time immune monitoring, could elicit persistent, antigen-specific T cells, and whether treatment was associated with changes in PSA doubling time (PSA DT). Sixteen patients with castration-resistant, nonmetastatic prostate cancer received six immunizations at 2-week intervals and then either quarterly (arm 1) or as determined by multiparameter immune monitoring (arm 2). Patients were on study a median of 16 months; four received 24 vaccinations. Only one event associated with treatment >grade 2 was observed. Six of 16 (38%) remained metastasis-free at 2 years. PAP-specific T cells were elicited in 12 of 16 (75%), predominantly of a Th1 phenotype, which persisted in frequency and phenotype for at least 1 year. IFNγ-secreting T-cell responses measured by ELISPOT were detectable in 5 of 13 individuals at 1 year, and this was not statistically different between study arms. The overall median fold change in PSA DT from pretreatment to posttreatment was 1.6 (range, 0.6-7.0; P = 0.036). Repetitive immunization with a plasmid DNA vaccine was safe and elicited Th1-biased antigen-specific T cells that persisted over time. Modifications in the immunization schedule based on real-time immune monitoring did not increase the frequency of patients developing effector and memory T-cell responses with this DNA vaccine.