EMAIL THIS PAGE TO A FRIEND
Molecular Pharmaceutics
Design and development of novel mitochondrial targeted nanocarriers, DQAsomes for curcumin inhalation.
PMID 24852198
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
08511
Curcumin, analytical standard
C21H20O6
00280590
Curcumin, primary pharmaceutical reference standard
C21H20O6
78246
Curcumin, matrix substance for MALDI-MS, ≥99.5% (HPLC)
C21H20O6
1151855
Curcumin, United States Pharmacopeia (USP) Reference Standard
C21H20O6
D3768
Dequalinium chloride hydrate, ≥95% (titration)
C30H40Cl2N4 · xH2O
M2933
MES hydrate, BioPerformance Certified, suitable for cell culture, ≥99.5%
C6H13NO4S · xH2O
M5287
MES hydrate, ≥99.5% (titration), pH 2.5-4.0 (0.5 M in H2O), BioXtra
C6H13NO4S · xH2O
69890
MES hydrate, BioUltra, ≥99.5% (T)
C6H13NO4S · xH2O
M8250
MES hydrate, ≥99.5% (titration)
C6H13NO4S · xH2O
PHG0003
MES hydrate, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C6H13NO4S · xH2O
69889
MES monohydrate, BioUltra, for molecular biology, ≥99.5% (T)
C6H13NO4S · H2O
69892
MES monohydrate, BioXtra, ≥99.0% (T)
C6H13NO4S · H2O
RES0113M-A7
MES monohydrate, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C6H13NO4S · H2O
82648
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
C16H10
18868
Pyrene, certified reference material, TraceCERT®
C16H10
571245
Pyrene, sublimed grade, 99%
C16H10
185515
Pyrene, 98%
C16H10
82650
Pyrene, purum, for fluorescence, ≥97.0% (GC)
C16H10
BCR177R
Pyrene, BCR® certified Reference Material
C16H10
40082
Pyrene, certified reference material, 1000 μg/mL in methanol
C16H10
48570
Pyrene, analytical standard
C16H10