EMAIL THIS PAGE TO A FRIEND
JAMA ophthalmology
Fundus autofluorescence in Tay-Sachs disease.
PMID 24852271
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G8397 Monosialoganglioside GM2 from bovine brain, ≥95% (TLC)