EMAIL THIS PAGE TO A FRIEND
Osteoarthritis and Cartilage / OARS, Osteoarthritis Research Society
Liver X Receptor activation delays chondrocyte hypertrophy during endochondral bone growth.
PMID 24852699
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74440
(−)-Nopol, purum, ≥98.0% (sum of enantiomers, GC)
C11H18O
HPA002924
Anti-CDKN1C antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV34285
Anti-CORO1A (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV38899
Anti-CORO1A (AB2) antibody produced in rabbit, IgG fraction of antiserum
AV38900
Anti-CORO1A (AB3) antibody produced in rabbit, IgG fraction of antiserum
AV34286
Anti-CORO1A antibody produced in rabbit, affinity isolated antibody
HPA051132
Anti-CORO1A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4200078
Anti-Coronin-1A (N-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody
SAB1300483
Anti-KIP2 (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
P0357 Anti-p57Kip2 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
PHR1553
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO
SAB5300325
Monoclonal Anti-CDKN1C antibody produced in mouse, clone 3E3, ascites fluid
WH0011151M1
Monoclonal Anti-CORO1A antibody produced in mouse, clone 4G10, purified immunoglobulin, buffered aqueous solution
D4551
N,N-Dimethylformamide, for molecular biology, ≥99%
C3H7NO
227056
N,N-Dimethylformamide, anhydrous, 99.8%
C3H7NO
648531
N,N-Dimethylformamide, for HPLC, ≥99.9%
C3H7NO
437573
N,N-Dimethylformamide, ACS reagent, ≥99.8%
C3H7NO
72438
N,N-Dimethylformamide, analytical standard
C3H7NO
209643
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Se
204307
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Se
229865
Selenium, powder, −100 mesh, 99.99% trace metals basis
Se
209651
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Se
HT-SE101
Selenium, pellets, < 5mm, ≥99.999%
Se
GF66395212
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Se