EMAIL THIS PAGE TO A FRIEND
Oncology research
Clove extract inhibits tumor growth and promotes cell cycle arrest and apoptosis.
PMID 24854101
Abstract
Cloves (Syzygium aromaticum) have been used as a traditional Chinese medicinal herb for thousands of years. Cloves possess antiseptic, antibacterial, antifungal, and antiviral properties, but their potential anticancer activity remains unknown. In this study, we investigated the in vitro and in vivo antitumor effects and biological mechanisms of ethyl acetate extract of cloves (EAEC) and the potential bioactive components responsible for its antitumor activity. The effects of EAEC on cell growth, cell cycle distribution, and apoptosis were investigated using human cancer cell lines. The molecular changes associated with the effects of EAEC were analyzed by Western blot and (qRT)-PCR analysis. The in vivo effect of EAEC and its bioactive component was investigated using the HT-29 tumor xenograft model. We identified oleanolic acid (OA) as one of the components of EAEC responsible for its antitumor activity. Both EAEC and OA display cytotoxicity against several human cancer cell lines. Interestingly, EAEC was superior to OA and the chemotherapeutic agent 5-fluorouracil at suppressing growth of colon tumor xenografts. EAEC promoted G0/G1 cell cycle arrest and induced apoptosis in a dose-dependent manner. Treatment with EAEC and OA selectively increased protein expression of p21(WAF1/Cip1) and γ-H2AX and downregulated expression of cell cycle-regulated proteins. Moreover, many of these changes were at the mRNA level, suggesting transcriptional regulation by EAEC treatment. Our results demonstrate that clove extract may represent a novel therapeutic herb for the treatment of colorectal cancer, and OA appears to be one of the bioactive components.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405470
Anti-BIRC5 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002830
Anti-BIRC5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2502110
Anti-E2F1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2103144
Anti-E2F1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2900055
Anti-EPHA4 (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
E6900 Anti-EphA4 antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB2107644
Anti-EPHA4 antibody produced in rabbit, affinity isolated antibody
SAB4300387
Anti-RB1 (Ab-780) antibody produced in rabbit, affinity isolated antibody
SAB4300385
Anti-RB1 (Ab-795) antibody produced in rabbit, affinity isolated antibody
SAB4300386
Anti-RB1 (Ab-807) antibody produced in rabbit, affinity isolated antibody
AV33212
Anti-RB1 antibody produced in rabbit, affinity isolated antibody
SAB4501460
Anti-Survivin antibody produced in rabbit, affinity isolated antibody
S8191
Anti-Survivin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
35995
Eugenol, PESTANAL®, analytical standard
C10H12O2
PHR1269
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material
C10H12O2
79891
Eugenol, certified reference material, TraceCERT®
C10H12O2
01050595
Eugenol, primary reference standard
C10H12O2
E5000000
Eugenol, European Pharmacopoeia (EP) Reference Standard
C10H12O2
WH0000332M1
Monoclonal Anti-BIRC5 antibody produced in mouse, clone 5B10, purified immunoglobulin, buffered aqueous solution
WH0001869M1
Monoclonal Anti-E2F1 antibody produced in mouse, clone 2E10, purified immunoglobulin, buffered aqueous solution
WH0002043M2
Monoclonal Anti-EPHA4 antibody produced in mouse, clone 6H7, purified immunoglobulin, buffered aqueous solution
SAB5300213 Monoclonal Anti-RB1 antibody produced in mouse, clone 7E4B8, ascites fluid
PLA0139
Rabbit anti-E2F1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
AV04006
Anti-E2F1 (AB1) antibody produced in rabbit, affinity isolated antibody
SAB2900054
Anti-EPHA4 (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500269
Anti-Survivin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB5300482 Monoclonal Anti-EPHA4 antibody produced in mouse, clone 7D3D4, ascites fluid
SAB5300483 Monoclonal Anti-EPHA4 antibody produced in mouse, clone 6C1B6, ascites fluid
SAB5300510 Monoclonal Anti-BIRC5 antibody produced in mouse, clone 2H5H2, ascites fluid
SAB5300511
Monoclonal Anti-BIRC5 antibody produced in mouse, clone 1H5, purified immunoglobulin, buffered aqueous solution