EMAIL THIS PAGE TO A FRIEND
Biology of Reproduction
MicroRNA-31 is significantly elevated in both human endometrium and serum during the window of implantation: a potential biomarker for optimum receptivity.
PMID 24855107