EMAIL THIS PAGE TO A FRIEND
Journal of bioscience and bioengineering
Biochemical characterization of Aspergillus oryzae native tannase and the recombinant enzyme expressed in Pichia pastoris.
PMID 24856589
Abstract
In this study, the biochemical properties of the recombinant tannase from Aspegillus oryzae were compared with those of the native enzyme. Extracellular native tannase was purified from a commercial enzyme source. Recombinant tannase highly expressed in Pichia pastoris was prepared as an active extracellular protein. Purified native and recombinant tannases produced smeared bands with apparent molecular masses of 45-80xa0kDa and 45-75xa0kDa, respectively, by sodium dodecyl sulfate-polyacrylamide gel electrophoresis. After N-deglycosylation, the native enzyme yielded molecular masses of 33xa0kDa and 30xa0kDa, whereas the recombinant enzyme yielded molecular masses of 34xa0kDa and 30xa0kDa. Purified native and recombinant tannases had an optimum pH of 4.0-5.0 and 5.0, respectively, and were stable up to 40°C. After N-deglycosylation, both enzymes exhibited reduced thermostability. Catalytic efficiencies of both purified enzymes were greater with natural substrates, such as (-)-catechin, (-)-epicatechin, and (-)-epigallocatechin gallates, than those with synthetic substrates, such as methyl, ethyl, and propyl gallates. However, there were no activities against the methyl esters of ferulic, p-coumaric, caffeic, and sinapic acids, which indicate feruloyl esterase activity, or the ethyl, propyl, and butyl esters of 4-hydroxybenzoic acid, which indicate paraben hydrolase activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42395 Tannase from Aspergillus ficuum, powder, white, ≥150 U/g