EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Two-photon neuronal and astrocytic stimulation with azobenzene-based photoswitches.
PMID 24857186
Abstract
Synthetic photochromic compounds can be designed to control a variety of proteins and their biochemical functions in living cells, but the high spatiotemporal precision and tissue penetration of two-photon stimulation have never been investigated in these molecules. Here we demonstrate two-photon excitation of azobenzene-based protein switches and versatile strategies to enhance their photochemical responses. This enables new applications to control the activation of neurons and astrocytes with cellular and subcellular resolution.