EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Metabolomics-based search for therapeutic agents for non-alcoholic steatohepatitis.
PMID 24857839
Abstract
Non-alcoholic fatty liver disease (NAFLD) is the commonest form of chronic liver disease in developed countries. Non-alcoholic steatohepatitis (NASH), which represents advanced stage NAFLD, is increasingly being recognized as a major cause of liver-related morbidity and mortality. However, no effective drugs against NASH have yet been developed. Therefore, we searched for candidate therapeutic agents based on the changes in levels of hepatic metabolites via gas chromatography mass spectrometry (GC/MS)-based metabolome analysis of livers from methionine-choline deficient (MCD) diet-fed mice, a mouse model of NASH. The metabolites were extracted from the livers of the MCD diet-fed mice and then analyzed using GC/MS. Subsequently, the MCD diet-fed mice were supplemented with hypotaurine, and the therapeutic effects of hypotaurine against steatohepatitis were evaluated. Ninety-nine metabolites were detected in the livers of the MCD diet-induced steatohepatitis model mice. Among these metabolites, hypotaurine exhibited the greatest decrease in its concentration in the mice. Supplementation with 2 mmol/kgBW hypotaurine attenuated liver injuries and fat accumulation caused by the MCD diet-induced steatohepatitis. Furthermore, 10 mmol/kgBW hypotaurine supplementation ameliorated fibrosis and oxidative stress induced by the MCD diet. The present metabolome analysis-based study demonstrated that hypotaurine is a novel candidate therapeutic agent for NASH.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1643361
Taurine, United States Pharmacopeia (USP) Reference Standard
C2H7NO3S