EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Down-regulation of ATBF1 activates STAT3 signaling via PIAS3 in pacing-induced HL-1 atrial myocytes.
PMID 24857987
Abstract
Atrial fibrillation (AF) is progressive and is the most common clinical arrhythmia. It is associated with inflammatory changes characterized by signal transducer and activator of transcription 3 (STAT3) signaling. A zinc finger homeobox 3 (ZFHX3, also named AT-motif binding factor 1, ATBF1) gene variant has been found in patients with AF. However, the mechanism by which the ATBF1 leads to inflammation in AF remains unknown. The aim of this study was to investigate whether tachypacing induces a decrease in ATBF1 expression and then activates STAT3 signaling via protein inhibitor of activated STAT3 (PIAS3). Atrial (HL-1 myocytes) cells were cultured in the presence of rapid electrical stimulations. In tachypaced HL-1 cells, we found that ATBF1 and PIAS3 protein levels were decreased, while the level of phosphorylated STAT3 (p-STAT3) was highly up-regulated compared with that of total STAT3. Knockdown of ATBF1 enhanced this trend, while the overexpression of ATBF1 had the opposite effect. A binary complex of ATBF1 and PIAS3 was formed and then the DNA-binding ability of activated STAT3 was enhanced in tachypaced HL-1 cells. These data indicate that tachypacing decreased ATBF1, leading to enhanced STAT3 DNA-binding activity due to the reduced formation of a binary complex of ATBF1 and PIAS3.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500484
Anti-PIAS3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV32762
Anti-PIAS3 antibody produced in rabbit, IgG fraction of antiserum
SAB4300708
Anti-STAT3 (Ab-705) antibody produced in rabbit, affinity isolated antibody
SAB4300327
Anti-STAT3 (Ab-727) antibody produced in rabbit, affinity isolated antibody
SAB2500993
Anti-STAT3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2104461
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
SAB2104912
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
SAB2102317
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
S5933 Anti-STAT3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA059353
Anti-ZFHX3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501946
Anti-ZFHX3 antibody produced in rabbit, affinity isolated antibody
WH0000463M1
Monoclonal Anti-ATBF1 antibody produced in mouse, clone 3B1, purified immunoglobulin, buffered aqueous solution
WH0010401M3
Monoclonal Anti-PIAS3 antibody produced in mouse, clone 4F12, purified immunoglobulin, buffered aqueous solution
P0117
Monoclonal Anti-PIAS3 antibody produced in mouse, ~2 mg/mL, clone PIA3, purified immunoglobulin, buffered aqueous solution
SAB1404414
Monoclonal Anti-STAT3 antibody produced in mouse, clone 1D11-2A11, purified immunoglobulin, buffered aqueous solution
WH0006774M2
Monoclonal Anti-STAT3 antibody produced in mouse, clone 4D6, purified immunoglobulin, buffered aqueous solution
AMAB90777
Monoclonal Anti-STAT3 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0492, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90776
Monoclonal Anti-STAT3 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0490, purified immunoglobulin, buffered aqueous glycerol solution
PLA0255
Rabbit anti-STAT3 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
SAB2104021
Anti-PIAS3 antibody produced in rabbit, affinity isolated antibody
SAB1406487
Anti-STAT3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution