EMAIL THIS PAGE TO A FRIEND
Analytical and Bioanalytical Chemistry
Highly hydrophilic and nonionic poly(2-vinyloxazoline)-grafted silica: a novel organic phase for high-selectivity hydrophilic interaction chromatography.
PMID 24858469