EMAIL THIS PAGE TO A FRIEND
Aquatic toxicology (Amsterdam, Netherlands)
Photosynthetic and molecular responses of the marine diatom Thalassiosira pseudonana to triphenyltin exposure.
PMID 24858899
Abstract
This study aimed to investigate the responses of the marine diatom Thalassiosira pseudonana upon waterborne exposure to triphenyltin chloride (TPTCl) through determining their photosynthetic response, growth performance, and expressions of genes and proteins. Based on the growth inhibition test, the 96-h IC50 (i.e., median inhibition concentration) was found to be 1.09 μg/L (95% confidence interval (CI): 0.89-1.34 μg/L). According to photosynthetic parameters, the 96-h EC50s (i.e., median effect concentrations) were estimated at 1.54 μg/L (95% CI: 1.40-1.69 μg/L) and 1.51 μg/L (95% CI: 1.44-1.58 μg/L) for the maximum quantum yield of photosystem II (PSII) photochemistry (ΦPo) and the effective quantum yield of photochemical energy conversion in PSII (Φ2), respectively. Non-photochemical quenching in the algae was increased at low concentrations of TPTCl (0.5-1.0 μg/L) but it decreased gradually when the TPTCl concentration further increased from 1.0 to 2.5 μg/L. Results of gene expressions showed that lipid metabolism related genes were not influenced by TPTCl at 0.5 or 1.0 μg/L, while silica shell formation genes were down-regulated at 0.5 μg/L. Photosynthesis related genes were up-regulated at 0.5 μg/L TPTCl but were down-regulated at 1.0 μg/L TPTCl. Proteomics analysis revealed that relatively less proteins could be detected after exposure to 1.0 μg/L TPTCl (only about 50-60 spots) compared with that observed in the 0.5μg/L TPTCl treatment and two control groups (each with about 290-300 protein spots). At 0.5 μg/L TPTCl, five proteins were differentially expressed when compared with the seawater control and solvent control, and most of these proteins are involved in defence function to protect the biological systems from reactive oxygen species that generated by TPTCl. These proteins include oxygen-evolving enhancer protein 1 precursor, fucoxanthin chlorophyll a/c protein - LI818 clade, and mitochondrial manganese superoxide dismutase, which can function to maintain the capacity of PSII and stabilize the photosynthesis efficiency as reflected by the unchanged ΦPo and Φ2 values at 0.5 μg/L TPTCl. In contrast, the excess toxicity that caused by TPTCl at the high concentration (1.0 μg/L TPTCl) might directly damage the proteins, inhibit their expression, and/or cause the suppression of metabolism as indicated by the down-regulation of most studied proteins and genes, which could ultimately inhibit the photosynthesis and growth of the algae. Overall, this study comprehensively elucidated the toxicity effects of TPT on T. pseudonana, and partially revealed the molecular toxic mechanisms and corresponding defence responses in this model algal species.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45492
Fentin chloride, PESTANAL®, analytical standard
C18H15ClSn