EMAIL THIS PAGE TO A FRIEND
Polish journal of pharmacology and pharmacy
Comparative studies of doxorubicin and 4'-epidoxorubicin effects on cardiovascular system in rabbits.
PMID 2485907
Abstract
4'-epidoxorubicin (4'-epiDXR) was compared with doxorubicin (DXR) for myocardial effects in rabbits. Histologically, both drugs when given for six weeks induced a high incidence of diffused myocardial damage. The myocardial damage was less evident in rabbits treated with 4'-epiDXR than DXR for 3 weeks. Functionally 4'-epiDXR like DXR significantly reduced cardiac index (CI), stroke index (SI) and mean arterial blood pressure (MAP).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0600000 Deslanoside, European Pharmacopoeia (EP) Reference Standard
C47H74O19