EMAIL THIS PAGE TO A FRIEND
Experimental hematology
Generation of induced pluripotent stem cells derived from primary and secondary myelofibrosis patient samples.
PMID 24859480
Abstract
Induced pluripotent stem cells (iPS) derived from disease cells are expected to provide a new experimental material, especially for diseases from which samples are difficult to obtain. In this study, we generated iPS from samples from patients with primary and secondary myelofibrosis. The primary myelofibrosis cells had chromosome 13q deletions, and the secondary myelofibrosis (SMF) cells had JAK2V617F mutations. The myelofibrosis patient cell-derived iPS (MF-iPS) were confirmed as possessing these parental disease-specific genomic markers. The capacity to form three germ layers was confirmed by teratoma assay. By co-culture with specific feeder cells and cytokines, MF-iPS can re-differentiate into blood progenitor cells and finally into megakaryocytes. We found that mRNA levels of interleukin-8, one of the candidate cytokines related to the pathogenesis of myelofibrosis, was elevated predominantly in megakaryocytes derived from MF-iPS. Because megakaryocytes from myelofibrosis clones are considered to produce critical mediators to proliferate fibroblasts in the bone marrow and iPS can provide differentiated cells abundantly, the disease-specific iPS we established should be a good research tool for this intractable disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6273
2-Propylpentanoic acid
C8H16O2
SRP4037 FGF-2 human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
SRP4134 IL-3 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
SRP6171
IL-3 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
SRP3090 IL-3 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
SRP3096 IL-6 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
I1646 Interleukin-3 human, IL-3, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
I7389 Interleukin-3 human, IL-3, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
I2786
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, Carrier Free
I1395
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
SRP3151 SCF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
W302600
Sodium citrate dihydrate, ≥99%, FG
C6H5Na3O7 · 2H2O
S4641
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%
C6H5Na3O7 · 2H2O
C7254
Sodium citrate tribasic dihydrate, ≥98%
C6H5Na3O7 · 2H2O
C8532
Sodium citrate tribasic dihydrate, for molecular biology, ≥99%
C6H5Na3O7 · 2H2O
C3434
Sodium citrate tribasic dihydrate, BioXtra, ≥99.0% (titration)
C6H5Na3O7 · 2H2O
71402
Sodium citrate tribasic dihydrate, BioUltra, for molecular biology, ≥99.5% (NT)
C6H5Na3O7 · 2H2O
71405
Sodium citrate tribasic dihydrate, puriss. p.a., ACS reagent, ≥99.0% (NT)
C6H5Na3O7 · 2H2O
71406
Sodium citrate tribasic dihydrate, purum p.a., ≥99.0% (NT)
C6H5Na3O7 · 2H2O
71409
Sodium citrate tribasic dihydrate, tested according to Ph Eur
C6H5Na3O7 · 2H2O
S7901 Stem Cell Factor human, SCF, recombinant, expressed in E. coli, powder, suitable for cell culture
S1804
Trisodium citrate dihydrate, meets USP testing specifications
C6H5Na3O7 · 2H2O
PHR1061
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C8H16O2
1708707
Valproic acid, United States Pharmacopeia (USP) Reference Standard
C8H16O2
V0033000
Valproic acid, European Pharmacopoeia (EP) Reference Standard
C8H16O2
Y0001422
Valproic acid for system suitability, European Pharmacopoeia (EP) Reference Standard
C8H16O2
V7259 Vascular Endothelial Growth Factor human, VEGF, recombinant, expressed in E. coli, powder, suitable for cell culture
SRP3182
VEGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested