EMAIL THIS PAGE TO A FRIEND
Acta Tropica
Detection of a broad range of Leishmania species and determination of parasite load of infected mouse by real-time PCR targeting the arginine permease gene AAP3.
PMID 24859532