EMAIL THIS PAGE TO A FRIEND
Acta neuropathologica
The autophagy/lysosome pathway is impaired in SCA7 patients and SCA7 knock-in mice.
PMID 24859968
Abstract
There is still no treatment for polyglutamine disorders, but clearance of mutant proteins might represent a potential therapeutic strategy. Autophagy, the major pathway for organelle and protein turnover, has been implicated in these diseases. To determine whether the autophagy/lysosome system contributes to the pathogenesis of spinocerebellar ataxia type 7 (SCA7), caused by expansion of a polyglutamine tract in the ataxin-7 protein, we looked for biochemical, histological and transcriptomic abnormalities in components of the autophagy/lysosome pathway in a knock-in mouse model of the disease, postmortem brain and peripheral blood mononuclear cells (PBMC) from patients. In the mouse model, mutant ataxin-7 accumulated in inclusions immunoreactive for the autophagy-associated proteins mTOR, beclin-1, p62 and ubiquitin. Atypical accumulations of the autophagosome/lysosome markers LC3, LAMP-1, LAMP2 and cathepsin-D were also found in the cerebellum of the SCA7 knock-in mice. In patients, abnormal accumulations of autophagy markers were detected in the cerebellum and cerebral cortex of patients, but not in the striatum that is spared in SCA7, suggesting that autophagy might be impaired by the selective accumulation of mutant ataxin-7. In vitro studies demonstrated that the autophagic flux was impaired in cells overexpressing full-length mutant ataxin-7. Interestingly, the expression of the early autophagy-associated gene ATG12 was increased in PBMC from SCA7 patients in correlation with disease severity. These results provide evidence that the autophagy/lysosome pathway is impaired in neurons undergoing degeneration in SCA7. Autophagy/lysosome-associated molecules might, therefore, be useful markers for monitoring the effects of potential therapeutic approaches using modulators of autophagy in SCA7 and other autophagy/lysosome-associated neurodegenerative disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV36578
Anti-ANXA3 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV36579
Anti-ANXA3 (AB2) antibody produced in rabbit, IgG fraction of antiserum
HPA013398
Anti-ANXA3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA013431
Anti-ANXA3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
A8104 Anti-Ataxin-7 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4421
Anti-ATG12 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4423
Anti-ATG12 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
A8731
Anti-Atg12 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA045444
Anti-ATG16L2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA007639
Anti-ATG7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104138
Anti-ATXN7 antibody produced in rabbit, affinity isolated antibody
PRS3627
Anti-LAMP-2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
L0668
Anti-LAMP2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA029100
Anti-LAMP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701987
Anti-LAMP2 antibody produced in rabbit
SAB2702058
Anti-LAMP2 antibody produced in rabbit
SAB1305638 ANTI-LC3 (APG8A) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
L8793
Anti-LC3A antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB1411514
Anti-MAP1LC3A antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300571
Anti-MAP1LC3A antibody produced in rabbit, affinity isolated antibody
SAB2101425
Anti-MAP1LC3A antibody produced in rabbit, affinity isolated antibody
HPA007649
Anti-MAP1LC3A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3501065
Anti-MAP1LC3B antibody produced in rabbit, affinity isolated antibody
R4779
Anti-Rab7 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA006964
Anti-RAB7A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411418
Anti-RAB7A antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4100184
Monoclonal Anti-BECN1 antibody produced in mouse, clone BECN1-B129, culture supernatant
SAB4100208
Monoclonal Anti-BECN1 antibody produced in mouse, clone BECN1-A129, culture supernatant
SAB5300513 Monoclonal Anti-BECN1 antibody produced in mouse, clone 2A4, ascites fluid
WH0002965M1
Monoclonal Anti-GTF2H1 antibody produced in mouse, clone 1F12-1B5, purified immunoglobulin, buffered aqueous solution