EMAIL THIS PAGE TO A FRIEND
Rapid communications in mass spectrometry : RCM
Guanidinated protein internal standard for immunoaffinity-liquid chromatography/tandem mass spectrometry quantitation of protein therapeutics.
PMID 24861599
Abstract
A protein internal standard (IS) is essential and superior to a peptide IS to achieve reproducible results in the quantitation of protein therapeutics using immunoaffinity-liquid chromatography/tandem mass spectrometry (LC/MS/MS). Guanidination has been used as a protein post-modification technique for more than half a century. A decade ago, the modification was applied to lysine-ending peptides to enhance their MALDI responses and peptide sequencing coverage. However, rarely has tryptic digestion of guanidinated proteins been investigated, likely due to the early conclusion that trypsin did not hydrolyze peptide bonds involving homoarginine in guanidinated proteins. In this study, the opposite was observed. Guanidinated lysine residues of proteins did not hinder the access of trypsin allowing for proteolytic digestion. Based on this observation, a new concept of internal standard, named Guanidinated Protein Internal Standard (GP-IS), was proposed for LC/MS/MS quantitation of protein therapeutics. The GP-IS is prepared by treating a portion of the therapeutic protein (analyte) with guanidine to convert arginine residues in the protein into homoarginine residues. After tryptic digestion, the GP-IS produces a series of homoarginine-ending peptides plus another series of arginine-ending peptides. One of the homoarginine-ending peptides, which corresponds to the analyte surrogate (lysine-ending) peptide, was chosen as a peptide internal standard (GP-PIS) for LC/MS/MS quantitation. Using this GP-IS approach, a sensitive and robust immunoaffinity-LC/MS/MS assay was developed and fully validated with a linearity range from 10 to 1000 ng/mL using 200 μL of human serum for the quantitation of an Astellas protein drug in clinical development. The proposed strategy allows LC/MS/MS to play an ever-increasing role in bioanalytical support for protein therapeutics development because of its capability of completely tracking all variations from the beginning to the end of sample analysis, easier preparation compared to isotope-labeled protein-IS, and greater flexibility for changing to alternate analyte surrogate peptides.