EMAIL THIS PAGE TO A FRIEND
Epilepsia
USL255 extended-release topiramate: dose-proportional pharmacokinetics and tolerability in healthy volunteers.
PMID 24861853
Abstract
Evaluate the pharmacokinetics (PK), safety, and tolerability of single doses of once-daily USL255, Qudexy XR (topiramate) extended-release capsules, over a wide dosing range. Two single-dose, phase I studies in healthy adults were used to evaluate the PK profile and maximum tolerated dose (MTD) of USL255 from 25-1,400 mg. Standard PK parameters assessed included area under the plasma concentration-time curve (AUC) and maximum plasma concentration (Cmax ). Dose proportionality, linearity, and intersubject and intrasubject variability (coefficient of variation [%CV]) of AUC and Cmax were evaluated. Investigator-reported adverse events (AEs) were obtained throughout the studies. After the initial increase in plasma concentration levels immediately following administration of USL255 25-1,400 mg, plasma topiramate concentration-time profiles were flat up to 24 h after dosing. AUC was dose proportional from 25-1,400 mg, and Cmax was dose proportional from 50-1,400 mg; both AUC and Cmax were linear across the entire dose range. Low intersubject and intrasubject %CV values were observed for AUC0-t , AUC0-∞ , and Cmax (intersubject %CV: 20.2, 19.6, and 22.4%, respectively; intrasubject %CV of dose-normalized mean values: 10.8, 8.2, and 13.2%, respectively). USL255 was generally safe and well tolerated with MTD established at 1,200 mg. These results demonstrate that USL255 provides consistent plasma topiramate exposure across an extended-dosing interval and predictable plasma topiramate concentrations over a wide dosing range. Overall, the favorable safety profile and consistency of exposure suggest once-daily USL255 can be a useful treatment option for patients with epilepsy.