EMAIL THIS PAGE TO A FRIEND
Clinical oral implants research
Biphasic β-TCP mixed with silicon increases bone formation in critical site defects in rabbit calvaria.
PMID 24863557
Abstract
The aim of this study was to assess the bone regeneration of critical size defects in rabbit calvarias filled with β-TCP doped with silicon. Twenty-one New Zealand rabbits were used in this study. Two critical size defects were created in the parietal bones. Three experimental groups were evaluated: Test A (HA/β-TCP granules alone), Test B (HA/β-TCP granules plus 3% silicon), Control (empty defect). The animals were sacrificed at 8 and 12 weeks. Evaluation was performed by μCT analysis and histomorphometry. μCT evaluation showed higher volume reduction in Test A group compared with Test B (P < 0.05). The Test B group showed the highest values for cortical closure and bone formation around the particles, followed by Test A and controls (P < 0.05). Within the limitations of this animal study, it can be concluded that HA/β-TCP plus 3% silicon increases bone formation in critical size defects in rabbit calvarias, and the incorporation of 3% silicon reduces the resorption rate of the HA/β-TCP granules.
Related Materials