EMAIL THIS PAGE TO A FRIEND
The Biochemical Journal
Discovery of InsP6-kinases as InsP6-dephosphorylating enzymes provides a new mechanism of cytosolic InsP6 degradation driven by the cellular ATP/ADP ratio.
PMID 24865181
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P55805
Purine, 98%
C5H4N4