EMAIL THIS PAGE TO A FRIEND
Molecular Imaging and Biology : MIB : The Official Publication of the Academy of Molecular Imaging
In vitro and in vivo characterization of two C-11-labeled pet tracers for vesicular acetylcholine transporter.
PMID 24865402