EMAIL THIS PAGE TO A FRIEND
The American journal of tropical medicine and hygiene
Cutaneous leishmaniasis in Cuban immigrants to Texas who traveled through the Darién Jungle, Panama.
PMID 24865687
Abstract
Cutaneous leishmaniasis is rarely seen in the United States. Four Cuban immigrants traveled along the same route at different times from Cuba to Ecuador, then northward, including through the Darién Jungle in Panama. These patients had chronic ulcerative non-healing skin lesions and were given a diagnosis of leishmaniasis.