EMAIL THIS PAGE TO A FRIEND
Journal of Dermatological Science
Combination of HDAC inhibitor MS-275 and IL-2 increased anti-tumor effect in a melanoma model via activated cytotoxic T cells.
PMID 24866536