EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Improved micromachined column design and fluidic interconnects for programmed high-temperature gas chromatography separations.
PMID 24866564
Abstract
This work focuses on the development and experimental evaluation of micromachined chromatographic columns for use in a commercial gas chromatography (GC) system. A vespel/graphite ferrule based compression sealing technique is presented using which leak-proof fluidic interconnection between the inlet tubing and the microchannel was achieved. This sealing technique enabled separation at temperatures up to 350°C on a μGC column. This paper reports the first high-temperature separations in microfabricated chromatographic columns at these temperatures. A 2m microfabricated column using a double Archimedean spiral design with a square cross-section of 100μm×100μm has been developed using silicon microfabrication techniques. The microfabricated column was benchmarked against a 2m 100μm diameter commercial column and the performance between the two columns was evaluated in tests performed under identical conditions. High temperature separations of simulated distillation (ASTM2887) and polycyclic aromatic hydrocarbons (EPA8310) were performed using the μGC column in temperature programmed mode. The demonstrated μGC column along with the high temperature fixture offers one more solution toward potentially realizing a portable μGC device for the detection of semi-volatile environmental pollutants and explosives without the thermal limitations reported to date with μGC columns using epoxy based interconnect technology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF37993595
Silicon, rod, 100mm, diameter 10.0mm, single crystal - random orientation, 100%
Si
GF64686998
Silicon, rod, 100mm, diameter 2.0mm, crystalline, 100%
Si
GF22929307
Silicon, rod, 100mm, diameter 5.0mm, crystalline, 100%
Si
GF16363391
Silicon, rod, 10mm, diameter 2.0mm, crystalline, 100%
Si
GF66465276
Silicon, rod, 25mm, diameter 2.0mm, crystalline, 100%
Si
GF43555664
Silicon, rod, 25mm, diameter 3.15mm, single crystal - random orientation, 100%
Si
GF70438518
Silicon, rod, 40mm, diameter 20mm, single crystal, -100, 99.999%
Si
GF35274113
Silicon, rod, 40mm, diameter 20mm, single crystal, -111, 99.999%
Si
GF05482852
Silicon, rod, 50mm, diameter 2.0mm, crystalline, 100%
Si
GF91515034
Silicon, rod, 50mm, diameter 3.15mm, single crystal - random orientation, 100%
Si
GF14076930
Silicon, rod, 50mm, diameter 5.0mm, crystalline, 100%
Si
GF28448691
Silicon, rod, 50mm, diameter 5mm, single crystal, -100, 99.999%
Si
GF40945754
Silicon, rod, 50mm, diameter 5mm, single crystal, -111, 99.999%
Si
GF64342188
Silicon, rod, 50mm, diameter 6mm, single crystal, -111, 99.999%
Si
GF10907377
Silicon, rod, 80mm, diameter 20mm, single crystal, -100, 99.999%
Si
GF91141792
Silicon, sheet, 10x10mm, thickness 0.5mm, single crystal, -100, 100%
Si
GF71817543
Silicon, sheet, 10x10mm, thickness 0.6mm, single crystal, -100, 100%
Si
GF75683212
Silicon, sheet, 14x14mm, thickness 1.0mm, single crystal, -100, 100%
Si
GF84840004
Silicon, sheet, 14x14mm, thickness 1.0mm, single crystal, -111, 100%
Si
GF20311254
Silicon, sheet, 25x25mm, thickness 1.0mm, polycrystalline, 99.999%
Si
GF41762493
Silicon, sheet, 25x25mm, thickness 1.0mm, single crystal, -100, 100%
Si
GF85621923
Silicon, sheet, 40x40mm, thickness 3.0mm, single crystal, p-type, 100%
Si
GF73529051
Silicon, sheet, 52x52mm, thickness 1.0mm, polycrystalline, 99.999%
Si
GF23955622
Silicon, single crystal, -100, n-type, phosphorus doped, diameter 150mm, length 0.65mm, 100%
Si
GF06693756
Silicon, single crystal, -111, n-type, phosphorus doped, diameter 76.2mm, length 0.50mm, 100%
Si
GF68832805
Silicon, single crystal, -111, n-type, phosphorus doped, diameter 76.2mm, length 0.75mm, 100%
Si
GF52781520
Silicon, sphere, 10pcs, diameter 2.0mm, precision sphere grade 25
Si
GF68982834
Silicon, sphere, 20pcs, diameter 2.0mm, precision sphere grade 25
Si
GF54682050
Silicon, sphere, 50pcs, diameter 2.0mm, precision sphere grade 25
Si
GF72027266
Silicon, disks, 13mm, thickness 0.38mm, single crystal, 100%
Si