EMAIL THIS PAGE TO A FRIEND
Journal of the American Society For Mass Spectrometry
Studying the chemistry of cationized triacylglycerols using electrospray ionization mass spectrometry and density functional theory computations.
PMID 24867430
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
66742
Dichloromethane, Selectophore, ≥99.5%
CH2Cl2
02575
Dichloromethane, analytical standard
CH2Cl2
48538
Dichloromethane, analytical standard, stabilized
CH2Cl2
48600
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
CH2Cl2
40042
Dichloromethane solution, certified reference material, 5000 μg/mL in methanol
CH2Cl2
M1550000
Methylene chloride, European Pharmacopoeia (EP) Reference Standard
CH2Cl2
PHR1557
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
CH2Cl2
330361
Soda ash, (99% as sodium carbonate or 58% as sodium oxide), technical grade, light
CNa2O3
791768
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
CNa2O3
S7795
Sodium carbonate, BioXtra, ≥99.0%
CNa2O3
204420
Sodium carbonate, 99.995% trace metals basis
CNa2O3
451614
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
CNa2O3
71345
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
CNa2O3
71363
Sodium carbonate, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
CNa2O3
28-2170
Sodium carbonate, JIS special grade, ≥99.5%
CNa2O3
72784
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
CNa2O3
56169
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
CNa2O3
490741
Sodium carbonate-12C, 99.9 atom % 12C
12CNa2O3
347817
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
CHF3O3S
158534
Trifluoromethanesulfonic acid, reagent grade, 98%
CHF3O3S
91738
Trifluoromethanesulfonic acid, purum, ≥98.0% (T)
CHF3O3S