EMAIL THIS PAGE TO A FRIEND
International journal of oncology
Modulation of uPA, MMPs and their inhibitors by a novel nutrient mixture in human glioblastoma cell lines.
PMID 24867464
Abstract
Brain tumors are highly aggressive tumors that are characterized by high levels of matrix metalloproteinase (MMP)-2 and -9 secretions that degrade the extracellular matrix (ECM) and basement membrane, allowing cancer cells to spread to distal organs. Proteases play a key role in tumor cell invasion and metastasis by digesting the basement membrane and ECM components. Strong clinical and experimental evidence demonstrates association of elevated levels of urokinase plasminogen activators (uPA) and MMPs with cancer progression, metastasis and shortened patient survival. MMP activities are regulated by specific tissue inhibitors of metalloproteinases (TIMPs). Our main objective was to study the effect of a nutrient mixture (NM) on the activity of uPA, MMPs and TIMPs in various human gliomas. Human glioblastoma (LN-18, T-98G and A-172) cell lines (ATCC) were cultured in their respective media and treated at confluence with NM at 0, 50, 100, 250, 500 and 1000 µg/ml. Analysis of uPA activity was carried out by fibrin zymography, MMPs by gelatinase zymography and TIMPs by reverse zymography. Glioblastoma cell lines LN-18 and T-98G expressed uPA, which was inhibited by NM in a dose-dependent manner. However, no bands corresponding to uPA were detected for the A-172 cell line. On gelatinase zymography, all three cell lines showed bands corresponding to MMP-2 and LN-18 and T-98G showed PMA (100 ng/ml)-induced MMP-9. NM inhibited their expression in a dose-dependent manner. Activity of TIMP-2 was upregulated by NM in all glioma cell lines in a dose-dependent manner. Analysis revealed a positive correlation between uPA and MMP-2 and a negative correlation between uPA/MMPs and TIMP-2. These findings suggest the therapeutic potential of NM in the treatment of gliomas.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1106
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
C6H14N4O2
23128
L-Lysine, analytical standard
C6H14N2O2
62850
L-Lysine solution, purum, 50% in H2O
C6H14N2O2
1568506
L-Proline, United States Pharmacopeia (USP) Reference Standard
C5H9NO2
93693
L-Proline, certified reference material, TraceCERT®
C5H9NO2
PHR1332
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
C5H9NO2
463728
Manganese, powder, ≥99.9% trace metals basis
Mn
266132
Manganese, powder, −325 mesh, ≥99% trace metals basis
Mn
266167
Manganese, chips, thickness <2.0 mm, 99%
Mn
GF87355334
Manganese, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
Mn
GF38495885
Manganese, rod, 50mm, diameter 4.0mm, cast, 99.5%
Mn
GF11021094
Manganese, rod, 10mm, diameter 4.0mm, cast, 99.5%
Mn
GF81243929
Manganese, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
Mn
GF83932768
Manganese, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
Mn
GF92898983
Manganese, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 98.7%
Mn
GF74177736
Manganese, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
Mn
GF99005193
Manganese, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 98.7%
Mn
GF90559794
Manganese, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 98.7%
Mn
GF03428625
Manganese, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
Mn
GF31519975
Manganese, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%
Mn
GF60677646
Manganese, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 98.7%
Mn
GF64815239
Manganese, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 98.7%
Mn
GF41418378
Manganese, foil, not light tested, 50x50mm, thickness 0.003mm, permanent polyester support, 98.7%
Mn
GF33640860
Manganese, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 98.7%
Mn
GF85367605
Manganese, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 98.7%
Mn
GF87981564
Manganese, foil, not light tested, 50x50mm, thickness 0.006mm, permanent polyester support, 98.7%
Mn
GF53022833
Manganese, foil, not light tested, 50x50mm, thickness 0.007mm, permanent polyester support, 98.7%
Mn
GF71274258
Manganese, foil, not light tested, 50x50mm, thickness 0.008mm, permanent polyester support, 98.7%
Mn
GF70055524
Manganese, foil, not light tested, 50x50mm, thickness 0.009mm, permanent polyester support, 98.7%
Mn
GF86610539
Manganese, foil, not light tested, 50x50mm, thickness 0.0125mm, permanent polyester support, 98.7%
Mn