EMAIL THIS PAGE TO A FRIEND
West African journal of medicine
Severe hypertension and haemospermia--a case report.
PMID 2486779
Abstract
Haemospermia is an uncommon disorder of diverse aetiology, which includes diseases of the urethra, prostate and hypertension. There is no report of haemospermia in this environment and this communication describes a case associated with severe hypertension.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H8760
Hydroflumethiazide, analytical standard, ~97%
C8H8F3N3O4S2