EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Antifungal resistance to fluconazole and echinocandins is not emerging in yeast isolates causing fungemia in a Spanish tertiary care center.
PMID 24867979
Abstract
Accurate knowledge of fungemia epidemiology requires identification of strains to the molecular level. Various studies have shown that the rate of resistance to fluconazole ranges from 2.5% to 9% in Candida spp. isolated from blood samples. However, trends in antifungal resistance have received little attention and have been studied only using CLSI M27-A3 methodology. We assessed the fungemia epidemiology in a large tertiary care institution in Madrid, Spain, by identifying isolates to the molecular level and performing antifungal susceptibility testing according to the updated breakpoints of European Committee for Antimicrobial Susceptibility Testing (EUCAST) definitive document (EDef) 7.2. We studied 613 isolates causing 598 episodes of fungemia in 544 patients admitted to our hospital (January 2007 to December 2013). Strains were identified after amplification and sequencing of the ITS1-5.8S-ITS2 region and further tested for in vitro susceptibility to amphotericin B, fluconazole, posaconazole, voriconazole, micafungin, and anidulafungin. Resistance was defined using EUCAST species-specific breakpoints, and epidemiological cutoff values (ECOFFs) were applied as tentative breakpoints. Most episodes were caused by Candida albicans (46%), Candida parapsilosis (28.7%), Candida glabrata (9.8%), and Candida tropicalis (8%). Molecular identification enabled us to better detect cryptic species of Candida guilliermondii and C. parapsilosis complexes and episodes of polyfungal fungemia. The overall percentage of fluconazole-resistant isolates was 5%, although it was higher in C. glabrata (8.6%) and non-Candida yeast isolates (47.4%). The rate of resistance to echinocandins was 4.4% and was mainly due to the presence of intrinsically resistant non-Candida species. Resistance mainly affected non-Candida yeasts. The rate of resistance to fluconazole and echinocandins did not change considerably during the study period.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000005 Amphotericin B, European Pharmacopoeia (EP) Reference Standard
C47H73NO17