EMAIL THIS PAGE TO A FRIEND
International Journal of Molecular Sciences
Quantitative proteomics to characterize specific histone H2A proteolysis in chronic lymphocytic leukemia and the myeloid THP-1 cell line.
PMID 24871368