EMAIL THIS PAGE TO A FRIEND
American Journal of Hypertension
Telmisartan exerts sustained blood pressure control and reduces blood pressure variability in metabolic syndrome by inhibiting sympathetic activity.
PMID 24871627