EMAIL THIS PAGE TO A FRIEND
FEBS letters
Dynamics and dispensability of variant-specific histone H1 Lys-26/Ser-27 and Thr-165 post-translational modifications.
PMID 24873882
Abstract
In mammals, the linker histone H1, involved in DNA packaging into chromatin, is represented by a family of variants. H1 tails undergo post-translational modifications (PTMs) that can be detected by mass spectrometry. We developed antibodies to analyze several of these as yet unexplored PTMs including the combination of H1.4 K26 acetylation or trimethylation and S27 phosphorylation. H1.2-T165 phosphorylation was detected at S and G2/M phases of the cell cycle and was dispensable for chromatin binding and cell proliferation; while the H1.4-K26 residue was essential for proper cell cycle progression. We conclude that histone H1 PTMs are dynamic over the cell cycle and that the recognition of modified lysines may be affected by phosphorylation of adjacent residues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-1372
Anti-acetyl (Lys26) phospho (Ser27) Histone H1.4 Antibody, Anti-acetyl (Lys26) phospho (Ser27) Histone H1.4 Antibody is a rabbit polyclonal antibody for detection of acetyl (Lys26) phospho (Ser27) Histone H1.4 also known as Histone H1.4, Histone H1b & has been validated in WB, DB.
06-1370
Anti-phospho-Histone H1 (Thr165) Antibody, Use Anti-phospho-Histone H1 (Thr165) Antibody (Rabbit Polyclonal Antibody) validated in WB, DB to detect phospho-Histone H1 (Thr165) also known as Histone H1.2, Histone H1d.
C3161
Calcium phosphate tribasic, BioReagent, plant cell culture tested, powder
HCa5O13P3
C5267
Calcium phosphate tribasic, 34.0-40.0% Ca basis
HCa5O13P3
702153
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Ca5HO13P3
H0252
Hydroxyapatite, Type I, buffered aqueous suspension
HCa5O13P3
55497
Hydroxyapatite, ′fast flow′
HCa5O13P3
04238
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
HCa5O13P3
574791
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
HCa5O13P3
21223
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
HCa5O13P3
289396
Hydroxyapatite, reagent grade, powder, synthetic
HCa5O13P3
677418
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
HCa5O13P3
693863
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
HCa5O13P3