EMAIL THIS PAGE TO A FRIEND
Nature Communications
PADI4 acts as a coactivator of Tal1 by counteracting repressive histone arginine methylation.
PMID 24874575
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09-818
Anti-CARM1 Antibody, Anti-CARM1 Antibody is a rabbit polyclonal antibody for detection of CARM1 also known as Protein arginine N-methyltransferase 4, coactivator-associated arginine methyltransferase 1 & has been validated in WB.
07-585
Anti-dimethyl-Histone H3 (Arg2) Antibody, Anti-dimethyl-Histone H3 (Arg2) Antibody is a Rabbit Polyclonal Antibody for detection of dimethyl-Histone H3 (Arg2) also known as H3R2me2, Histone H3 (di methyl R2) & has been validated in WB, PIA.
04-123
Anti-TAL-1 Antibody, clone BTL73, Use Anti-TAL-1 Antibody, clone BTL73 (Mouse Monoclonal Antibody) validated in WB, IHC to detect TAL-1 also known as SCL, T-cell acute lymphocytic leukemia 1, TCL5, TAL-1, Stem cell protein.
AV38820
Anti-CTCF (AB1) antibody produced in rabbit, affinity isolated antibody
AV100757
Anti-CTCF (AB2) antibody produced in rabbit, affinity isolated antibody
HPA004122
Anti-CTCF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
49897
Formate standard for IC, 1.000 g/L in H2O, analytical standard
CH2O2
06473
Formic acid solution, BioUltra, 1.0 M in H2O
CH2O2
AMAB90666
Monoclonal Anti-CTCF antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0307, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90664
Monoclonal Anti-CTCF antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0305, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90663
Monoclonal Anti-CTCF antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0304, purified immunoglobulin, buffered aqueous glycerol solution
SAB5300120 Monoclonal Anti-CTCF antibody produced in mouse, clone 1D11, ascites fluid