EMAIL THIS PAGE TO A FRIEND
Bioresource Technology
Operational stability of naringinase PVA lens-shaped microparticles in batch stirred reactors and mini packed bed reactors-one step closer to industry.
PMID 24874877