EMAIL THIS PAGE TO A FRIEND
Bioorganic Chemistry
Design and synthesis of marine natural product-based 1H-indole-2,3-dione scaffold as a new antifouling/antibacterial agent against fouling bacteria.
PMID 24875126