EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Chronic haloperidol increases voltage-gated Na+ currents in mouse cortical neurons.
PMID 24875357
Abstract
Typical antipsychotics are characterized by extrapyramidal syndrome (EPS). Previous studies demonstrated that typical antipsychotics could inhibit neuronal voltage-gated sodium channel (VGSC). However, EPS typically emerge only upon prolonged exposure. As a result, we examined effects of haloperidol, a prototype typical antipsychotic, on neuronal VGSC upon incubation for varying duration. Briefly, VGSC currents were activated and recorded using a whole-cell patch-clamp technique in primary culture of mouse cortical neurons. VGSC activity was inhibited by acute haloperidol exposure (for minutes), but enhanced in a time- and concentration-dependent manner by chronic haloperidol exposure (for hours). The effects of chronic haloperidol were associated with increased expression of VGSC subunits as well as corresponding electrophysiological channel properties. In summary, we found enhanced VGSC currents upon chronic haloperidol exposure in cortical neurons in contrast to inhibition by acute haloperidol exposure. Such a results may contribute to EPS of typical antipsychotics.