EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
Quantification of folate metabolites in serum using ultraperformance liquid chromatography tandem mass spectrometry.
PMID 24878879
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
44925
DL-Homocysteine, analytical standard
C4H9NO2S
A9384
S-(5′-Adenosyl)-L-homocysteine, crystalline
C14H20N6O5S