EMAIL THIS PAGE TO A FRIEND
Transplantation
Heat shock protein-27 delays acute rejection after cardiac transplantation: an experimental model.
PMID 24879379
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1086
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin, ≥97% (HPLC), powder
C30H37N5O13